(±)-Isonipecotic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A11795-14 A11795-22
CAAAA11795-14EA 82.63 CAD
CAAAA11795-14 CAAAA11795-22
(±)-Isonipecotic acid 98%
(±)-Isonipecotic acid

A GABA-A receptor partial agonist


Purity: 98% CAS No.: 498-94-2 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Odor: Odorless Melting Point: 350° Beilstein Reference: 112553 Merck Reference: 14,5189 MDL No.: MFCD00006004

Formula: C₆H₁₁NO₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: >350 °C
Storage Temperature: Ambient
MDL Number: MFCD00006004
CAS Number: 498-94-2
EINECS: 207-872-3
Merck Index: 14,5189

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR