(2,4-Dichlorophenyl)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
DCAA, 2,4-Dichlorophenylacetic acid
A11791-22
A11791-06
A11791-14
CAAAAA11791-22EA
312.84
CAD
CAAAAA11791-22
CAAAA11791-06
CAAAA11791-14
(2,4-Dichlorophenyl)acetic acid 98+%
(2,4-Dichlorophenyl)acetic acid
MDL: MFCD00004318
Beilstein Registry No.: 1948767
Purity: 98+%
CAS No.: 19719-28-9
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Odor: Odorless
Melting Point: 128-130°
Beilstein Reference: 1948767
MDL No.: MFCD00004318
Formula:
(C₆H₃Cl₂)CH₂COOH MW: 205.04 g/mol Boiling Pt: 316 °C (1013 hPa) Melting Pt: 128…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004318 CAS Number: 19719-28-9 EINECS: 243-248-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...