Tetraethylammonium iodide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TEAI, Tetrylammonium iodide

A11783-0B A11783-18 A11783-30
CAAAA11783-0BEA 1633.72 CAD
CAAAA11783-0B CAAAA11783-18 CAAAA11783-30
Tetraethylammonium iodide 98+%
Tetraethylammonium iodide

MDL: MFCD00011829 Beilstein Registry No.: 3562649 Fieser: 12,465


Purity: 98+% CAS No.: 68-05-3 Molecular Formula: (CH3CH2)4NI Formula Weight: 257.17 Melting Point: ca 320° dec. Beilstein Reference: 3562649

Formula: C₈H₂₀IN
MW: 257.16 g/mol
Melting Pt: ∼ 320 °C (dec.)
Density: 1.566 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011829
CAS Number: 68-05-3
EINECS: 200-676-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR