2-Nonanone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11781-22
A11781-0B
A11781-18
CAAAAA11781-22EA
78.05
CAD
CAAAAA11781-22
CAAAA11781-0B
CAAAA11781-18
2-Nonanone 98+%
2-Nonanone
MDL: MFCD00009553
Beilstein Registry No.: 1743645
Purity: 98+%
CAS No.: 821-55-6
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 64°(147°F)
Refractive Index: 1.4210
Melting Point: -15°
Boiling Point: 195°
Beilstein Reference: 1743645
MDL No.: MFCD00009553
Formula:
C₉H₁₈O Boiling Pt: 195 °C Melting Pt: –15 °C Density: 0.826 Flash Pt: 64 °C(147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009553 CAS Number: 821-55-6 EINECS: 212-480-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...