2-Mesitylenesulfonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4,6-Trimethylbenzenesulphonyl chloride, 2,4,6-Trimethylbenzenesulfonyl chloride, Mesitylene-2-sulphonyl chloride, Mesitylene-2-sulfonyl chloride

A11775-14 A11775-22 A11775-36
CAAAA11775-14EA 102.54 CAD
CAAAA11775-14 CAAAA11775-22 CAAAA11775-36
2-Mesitylenesulfonyl chloride 99%
2-Mesitylenesulphonyl chloride

MDL: MFCD00007434 Beilstein Registry No.: 1107601 Fieser: 1,661 5,430 6,625 8,318


Purity: 99% CAS No.: 773-64-8 Molecular Formula: C9H11ClO2S Formula Weight: 218.70 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Odor: Irritating Melting Point: 53-57° Boiling Point: 150°/20mm Beilstein Reference: 1107601 UN No.: 3261 MDL No.: MFCD00007434

Formula: C₉H₁₁ClO₂S
Boiling Pt: 150 °C/20 mm
Melting Pt: 53…57 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007434
CAS Number: 773-64-8
EINECS: 212-257-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR