3-Iodo-o-xylene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dimethyl-3-iodobenzene, 2,3-Dimethyl-1-iodobenzene, 1-Iodo-2,3-dimethylbenzene, 2,3-Dimethyliodobenzene

A11774-06 A11774-14
CAAAAA11774-22EA 1564.72 CAD
CAAAAA11774-22 CAAAA11774-06 CAAAA11774-14
3-Iodo-o-xylene 97%
3-Iodo-o-xylene

MDL: MFCD00041437 Beilstein Registry No.: 1929733

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 98…100 °C/15 mm
Density: 1.640
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041437
CAS Number: 31599-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR