Allyl bromide 99% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromopropene, 3-Propenyl bromide

Stabilization: 300-1000 ppm Stabilised with propylene oxide

A11766-0B A11766-30
CAAAA11766-0BEA 262.28 CAD
CAAAA11766-0B CAAAA11766-30
Allyl bromide 99% stabilised
Allyl bromide

MDL: MFCD00000244


Purity: 99% CAS No.: 106-95-6 Molecular Formula: C3H5Br Formula Weight: 120.98 Flash Point: -2°(28°F) Physical Form: Liquid Density: 1.398 Refractive Index: 1.4700 Sensitivity: Light Sensitive Appearance: Colorless Odor: Pungent Melting Point: -119° Boiling Point: 70-71° Merck Reference: 14,288 UN No.: 1099 MDL No.: MFCD00000244

Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 70
Melting Pt: -119
Density: 1.398
Flash Pt: –2 °C (28 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000244
CAS Number: 106-95-6
EINECS: 203-446-6
UN: 1099
ADR: 3 I
Merck Index: 14,288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR