Riboflavine (vitamin B2) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

A11764-14 A11764-22
CAAAA11764-14EA 64.91 CAD
CAAAA11764-14 CAAAA11764-22
Riboflavine (vitamin B2) 98%
Riboflavine (Vitamin B2)

Photoinitiator of polymerization for PAGE gels


Purity: 98% CAS No.: 83-88-5 Molecular Formula: C17H20N4O6 Formula Weight: 376.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Odor: Odorless Melting Point: 290° Merck Reference: 14,8200 MDL No.: MFCD00005022


Photoinitiator of polymerization for PAGE gels

Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 14,8200

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR