9,10-Phenanthrenequinone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11762-06
A11762-14
A11762-22
CAAAAA11762-06EA
47.24
CAD
CAAAAA11762-06
CAAAA11762-14
CAAAA11762-22
9,10-Phenanthrenequinone 95%
9,10-Phenanthrenequinone
MDL: MFCD00001163
Beilstein Registry No.: 608838
Fieser: 1,827 2,317
Purity: 95%
CAS No.: 84-11-7
Molecular Formula: C14H8O2
Formula Weight: 208.22
Flash Point: 245°(473°F)
Melting Point: 207-212°
Boiling Point:
Formula:
C₁₄H₈O₂ Boiling Pt: >360 °C Melting Pt: 207…212 °C Density: 1.405 Flash Pt: 245 °C (473 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001163 CAS Number: 84-11-7 EINECS: 201-515-5 UN: 3077 ADR: 9,III Merck Index: 14,7213 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...