(±)-δ-Decalactone 97%

Supplier: Thermo Scientific Chemicals

A11756-14 A11756-22 A11756-36
CAAAA11756-14EA 125.93 CAD
CAAAA11756-14 CAAAA11756-22 CAAAA11756-36
(±)-δ-Decalactone 97%
(±)-δ-Decalactone

MDL: MFCD00006649 Beilstein Registry No.: 117520


Purity: 97% CAS No.: 705-86-2 Molecular Formula: C10H18O2 Formula Weight: 170.25 Flash Point: 145°(293°F) Refractive Index: 1.4580 Melting Point: -25° Boiling Point: 117-120°/0.02m Beilstein Reference: 117520 MDL No.: MFCD00006649

Formula: C₁₀H₁₈O₂
Boiling Pt: 117…120 °C/0.02 mm
Melting Pt: –25 °C
Density: 0.971
Flash Pt: 145 °C(293 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006649
CAS Number: 705-86-2
EINECS: 211-889-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR