(4-Chlorobutyl)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11753-03
A11753-06
A11753-14
CAAAA11753-03EA
59.88
CAD
CAAAA11753-03
CAAAA11753-06
CAAAA11753-14
(4-Chlorobutyl)benzene 97%
(4-Chlorobutyl)benzene
MDL: MFCD00037117
Beilstein Registry No.: 2042123
Purity: 97%
CAS No.: 4830-93-7
Molecular Formula: C6H5(CH2)4Cl
Formula Weight: 168.67
Flash Point: 104°(219°F)
Refractive Index: 1.5180
Boiling Point: 125-128°/20mm
Beilstein Reference: 2042123
MDL No.: MFCD00037117
Formula:
(C₆H₅)(CH₂)₄Cl Boiling Pt: 125…128 °C/20 mm Density: 1.026 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037117 CAS Number: 4830-93-7 EINECS: 225-408-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...