Pyridinium chlorochromate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PCC

A11752-14 A11752-22 A11752-36
CAAAAA11752-14EA 46.93 CAD
CAAAAA11752-14 CAAAA11752-22 CAAAA11752-36
Pyridinium chlorochromate 98%
Pyridinium chlorochromate

MDL: MFCD00013106 Fieser: 6,498 9,397 10,334 11,450 12,417 13,262 14,269 15,276 18,302


Purity: 98% CAS No.: 26299-14-9 Molecular Formula: C5H6ClCrNO3 Formula Weight: 215.56 Melting Point: ca 206° dec. Merck Reference: 14,7974 MDL No.: MFCD00013106

Formula: C₅H₆ClCrNO₃
MW: 215.56 g/mol
Melting Pt: ∼ 206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013106
CAS Number: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,III
Merck Index: 14,7974

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR