2-Bromo-4-fluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11751-06
A11751-14
CAAAA11751-06EA
81.37
CAD
CAAAA11751-06
CAAAA11751-14
CAAAA11751-22
2-Bromo-4-fluoroaniline 98%
2-Bromo-4-fluoroaniline
MDL: MFCD00042462
Beilstein Registry No.: 2802562
Purity: 98%
CAS No.: 1003-98-1
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 104°(219°F)
Refractive Index: 1.5830
Boiling Point: 221°
Beilstein Reference: 2802562
UN No.: 2810
MDL No.: MFCD00042462
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 221 °C Melting Pt: 41 Density: 1.670 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042462 CAS Number: 1003-98-1 UN: 2810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...