4-Cholesten-3-one 98+%
Supplier: Thermo Scientific Chemicals
A11743-06
A11743-14
CAAAA11743-06EA
137.46
CAD
CAAAA11743-06
CAAAA11743-14
4-Cholesten-3-one 98+%
4-Cholesten-3-one
MDL: MFCD00003663
Beilstein Registry No.: 4707774
Optical Rotation: +93°C (c=1 in chloroform)
Purity: 98+%
CAS No.: 601-57-0
Molecular Formula: C27H44O
Formula Weight: 384.65
Melting Point: 79-81°
Beilstein Reference: 4707774
Formula:
C₂₇H₄₄O Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003663 CAS Number: 601-57-0 EINECS: 210-005-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...