trans-2-Hydroxycinnamic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11740-14
A11740-22
CAAAA11740-14EA
141.25
CAD
CAAAA11740-14
CAAAA11740-22
CAAAA11740-36
trans-2-Hydroxycinnamic acid 98+%
trans-2-Hydroxycinnamic acid
MDL: MFCD00004379
Beilstein Registry No.: 1100900
Fieser: 20,192
Purity: 98+%
CAS No.: 614-60-8
Molecular Formula: C9H8O3
Formula Weight: 164.16
Melting Point: ca 210° dec.
Beilstein Reference: 1100900
Formula:
C₉H₈O₃ Boiling Pt: 330 °C (1013 hPa) Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004379 CAS Number: 614-60-8 EINECS: 210-386-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...