4'-Methoxybutyrophenone 97%

Supplier: Thermo Scientific Chemicals

A11737-03 A11737-06 A11737-14
CAAAA11737-03EA 45.35 CAD
CAAAA11737-03 CAAAA11737-06 CAAAA11737-14
4'-Methoxybutyrophenone 97%
4'-Methoxybutyrophenone

MDL: MFCD00027138 Beilstein Registry No.: 908286


Purity: 97% CAS No.: 4160-51-4 Molecular Formula: C11H14O2 Formula Weight: 178.23 Refractive Index: 1.5390 Boiling Point: 124-128°/4mm MDL No.: MFCD00027138

Formula: C₁₁H₁₄O₂
Boiling Pt: 124…128 °C (4 mmHg)
Melting Pt: 26 °C
Density: 1,0451 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00027138
CAS Number: 4160-51-4
EINECS: 223-995-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR