3'-Fluoroacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11733-06
A11733-14
CAAAA11733-06EA
93.85
CAD
CAAAA11733-06
CAAAA11733-14
CAAAA11733-22
3'-Fluoroacetophenone 98+%
3'-Fluoroacetophenone
MDL: MFCD00000338
Beilstein Registry No.: 907041
Purity: 98+%
CAS No.: 455-36-7
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 80°(176°F)
Refractive Index: 1.5090
Melting Point: -3°
Boiling Point: 80-81°/9mm
Beilstein Reference: 907041
MDL No.: MFCD00000338
Formula:
C₈H₇FO Boiling Pt: 80…81 °C/9 mm Melting Pt: –3 °C Density: 1,130 Flash Pt: 80 °C(176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000338 CAS Number: 455-36-7 EINECS: 207-245-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...