6-Nitro-m-cresol 97%

Supplier: Thermo Scientific Chemicals
Warning

A11729-14 A11729-22 A11729-36
CAAAA11729-14EA 79.63 CAD
CAAAA11729-14 CAAAA11729-22 CAAAA11729-36
6-Nitro-m-cresol 97%
6-Nitro-m-cresol

MDL: MFCD00007111 Beilstein Registry No.: 1868030


Purity: 97% CAS No.: 700-38-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 109°(228°F) Odor: Phenol-like Beilstein Reference: 1868030 UN No.: 2446 MDL No.: MFCD00007111

Formula: C₇H₇NO₃
Melting Pt: 53…56 °C
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007111
CAS Number: 700-38-9
EINECS: 211-843-0
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR