(Methylsulfonyl)benzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl phenyl sulphone, Methyl phenyl sulfone

A11727-09 A11727-18
CAAAA11727-09EA 120.87 CAD
CAAAA11727-09 CAAAA11727-18 CAAAA11727-30
(Methylsulfonyl)benzene 98+%
(Methylsulphonyl)benzene

MDL: MFCD00014741 Beilstein Registry No.: 1906914 Fieser: 6,43


Purity: 98+% CAS No.: 3112-85-4 Molecular Formula: C7H8O2S Formula Weight: 156.20 Melting Point: 85-88° Beilstein Reference: 1906914

Formula: C₇H₈O₂S
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00014741
CAS Number: 3112-85-4
EINECS: 221-476-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR