Syringic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

A11725-14 A11725-22
CAAAA11725-14EA 142.99 CAD
CAAAA11725-14 CAAAA11725-22
Syringic acid 98+%
Syringic acid

MDL: MFCD00002552 Beilstein Registry No.: 2115262


Purity: 98+% CAS No.: 530-57-4 Molecular Formula: C9H10O5 Formula Weight: 198.17 Melting Point: 206-208° Boiling Point: 192-193°/14mm MDL No.: MFCD00002552

Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 192…193 °C (14 mmHg)
Melting Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4
EINECS: 208-486-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR