Diethylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11716-AE
A11716-0F
A11716-AP
CAAAAA11716-AEEA
50.5
CAD
CAAAAA11716-AE
CAAAA11716-0F
CAAAA11716-AP
Diethylamine 99+%
Diethylamine
MDL: MFCD00009032
Beilstein Registry No.: 605268
Fieser: 4,148
Purity: 99+%
CAS No.: 109-89-7
Molecular Formula: C4H11N
Formula Weight: 73.14
Flash Point: -28°(-18°F)
Refractive Index: 1.3850
Odor: Ammonia-like
Melting Point: -50°
Boiling Point: 54-55°
Beilstein Reference: 605268
Merck Reference: 14,3111
UN No.: 1154
MDL No.: MFCD00009032
Formula:
(C₂H₅)₂NH Boiling Pt: 54…55 °C Melting Pt: –50 °C Density: 0.706 Flash Pt: –28 °C (–18 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009032 CAS Number: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3,II Merck Index: 14,3111 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...