Triphenylmethane 98%
Supplier: Thermo Scientific Chemicals
A11710-14
A11710-22
A11710-36
CAAAA11710-14EA
55.3
CAD
CAAAA11710-14
CAAAA11710-22
CAAAA11710-36
Triphenylmethane 98%
Triphenylmethane
MDL: MFCD00004763
Beilstein Registry No.: 1909753
Purity: 98%
CAS No.: 519-73-3
Molecular Formula: (C6H5)3CH
Formula Weight: 244.34
Melting Point: 92-94°
Boiling Point: 358-359°
Merck Reference: 14,9741
MDL No.: MFCD00004763
Formula:
C₁₉H₁₆ Boiling Pt: 358…359 °C Melting Pt: 92…94 °C Density: 1.010 Storage Temperature: Ambient |
MDL Number:
MFCD00004763 CAS Number: 519-73-3 EINECS: 208-275-0 Merck Index: 14,9741 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...