1-Fluoro-3-iodobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoroiodobenzene
A11706-14
A11706-22
CAAAAA11706-06EA
494.21
CAD
CAAAAA11706-06
CAAAA11706-14
CAAAA11706-22
1-Fluoro-3-iodobenzene 99%
1-Fluoro-3-iodobenzene
MDL: MFCD00001044
Beilstein Registry No.: 2039305
Purity: 99%
CAS No.: 1121-86-4
Molecular Formula: C6H4FI
Formula Weight: 222.00
Flash Point: 67°(152°F)
Refractive Index: 1.5840
Melting Point: -42°
Boiling Point: 77-78°/19mm
Beilstein Reference: 2039305
MDL No.: MFCD00001044
Formula:
C₆H₄FI MW: 222 g/mol Boiling Pt: 77…78 °C/19 mm Melting Pt: –42 °C Density: 1,898 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001044 CAS Number: 1121-86-4 EINECS: 214-339-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...