2-Chloro-2-methylbutane 95%

Supplier: Thermo Scientific Chemicals
Danger

A11703-06 A11703-14 A11703-22
CAAAA11703-06EA 50.09 CAD
CAAAA11703-06 CAAAA11703-14 CAAAA11703-22
2-Chloro-2-methylbutane 95%
2-Chloro-2-methylbutane

MDL: MFCD00000818 Beilstein Registry No.: 1696930


Purity: 95% CAS No.: 594-36-5 Molecular Formula: C5H11Cl Formula Weight: 106.60 Flash Point: -10°(14°F) Refractive Index: 1.4042 Melting Point: -73° Boiling Point: 85-86° Beilstein Reference: 1696930 UN No.: 1107 MDL No.: MFCD00000818

Formula: C₅H₁₁Cl
Boiling Pt: 85…86 °C
Melting Pt: –73 °C
Density: 0.866
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000818
CAS Number: 594-36-5
EINECS: 209-836-2
UN: 1107
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR