5-Nitroisophthalic acid 98%
Supplier: Thermo Scientific Chemicals
A11696-22
A11696-36
CAAAA11696-22EA
43.13
CAD
CAAAA11696-22
CAAAA11696-36
5-Nitroisophthalic acid 98%
5-Nitroisophthalic acid
MDL: MFCD00007254
Beilstein Registry No.: 1976587
Purity: 98%
CAS No.: 618-88-2
Molecular Formula: C8H5NO6
Formula Weight: 211.13
Odor: Odorless
Melting Point: 260-264°
Beilstein Reference: 1976587
MDL No.: MFCD00007254
Formula:
C₈H₅NO₆ Melting Pt: 260…264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007254 CAS Number: 618-88-2 EINECS: 210-568-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...