2-Bromoanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-methoxybenzene

A11692-14 A11692-22
CAAAA11692-14EA 69.99 CAD
CAAAA11692-14 CAAAA11692-22 CAAAA11692-36
2-Bromoanisole 98%
2-Bromoanisole

MDL: MFCD00000064 Beilstein Registry No.: 1859996


Purity: 98% CAS No.: 578-57-4 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 96°(204°F) Refractive Index: 1.5740 Melting Point: 2-3° Boiling Point: 220-222° Beilstein Reference: 1859996 MDL No.: MFCD00000064

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 220…222 °C
Melting Pt: 2…3 °C
Density: 1.511
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000064
CAS Number: 578-57-4
EINECS: 209-425-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR