1-Bromo-2-nitrobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11686-0B
A11686-18
A11686-30
CAAAA11686-0BEA
1567.36
CAD
CAAAA11686-0B
CAAAA11686-18
CAAAA11686-30
1-Bromo-2-nitrobenzene 99%
1-Bromo-2-nitrobenzene
MDL: MFCD00007045
Beilstein Registry No.: 2044109
Purity: 99%
CAS No.: 577-19-5
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Flash Point: 112°(233°F)
Melting Point: 40-44°
Boiling Point: 261°
Beilstein Reference: 2044109
UN No.: 3459
MDL No.: MFCD00007045
Formula:
C₆H₄BrNO₂ Boiling Pt: 261 °C Melting Pt: 40…44 °C Density: 1.62 g/cm³ (20 °C) Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007045 CAS Number: 577-19-5 EINECS: 209-409-0 UN: 3459 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...