2,4,6-Trichlorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11681-0B
A11681-30
CAAAA11681-0BEA
202.24
CAD
CAAAA11681-0B
CAAAA11681-0I
CAAAA11681-30
2,4,6-Trichlorophenol 98%
2,4,6-Trichlorophenol
MDL: MFCD00002172
Beilstein Registry No.: 776729
Purity: 98%
CAS No.: 88-06-2
Molecular Formula: C6H3Cl3O
Formula Weight: 197.45
Flash Point: 99°(210°F)
Physical Form: Cryst solid
Density: 1.675
Appearance: White to pale brown
Odor: Phenol-like
Melting Point: 63-68°
Boiling Point: 245-246°
Beilstein Reference: 776729
Merck Reference: 14,9644
UN No.: 2020
MDL No.: MFCD00002172
Formula:
C₆H₃Cl₃O MW: 197.45 g/mol Boiling Pt: 245…246 °C Melting Pt: 63…68 °C Density: 1.675 Flash Pt: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002172 CAS Number: 88-06-2 EINECS: 201-795-9 UN: 2020 ADR: 6.1 III Merck Index: 14,9644 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...