2-Nitro-p-anisidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11673-0E
A11673-22
A11673-36
CAAAA11673-0EEA
753.66
CAD
CAAAA11673-0E
CAAAA11673-22
CAAAA11673-36
2-Nitro-p-anisidine 99%
2-Nitro-p-anisidine
MDL: MFCD00007152
Beilstein Registry No.: 880318
Purity: 99%
CAS No.: 96-96-8
Molecular Formula: C7H8N2O3
Formula Weight: 168.15
Melting Point: 126-128°
Beilstein Reference: 880318
UN No.: 2811
MDL No.: MFCD00007152
Formula:
C₇H₈N₂O₃ Boiling Pt: ? 250 °C (1013 hPa) Melting Pt: 126…128 °C Flash Pt: ≥ 200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007152 CAS Number: 96-96-8 EINECS: 202-547-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...