Dibutylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11671-0F
A11671-AP
CAAAA11671-0FEA
235.97
CAD
CAAAA11671-0F
CAAAA11671-AP
Dibutylamine 98+%
Dibutylamine
MDL: MFCD00009429
Beilstein Registry No.: 506001
Fieser: 1,209
Purity: 98+%
CAS No.: 111-92-2
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 39°(102°F)
Physical Form: Clear liquid
Density: 0.767
Refractive Index: 1.4169
Appearance: Colorless
Odor: Amine-like
Melting Point: -62°
Boiling Point: 159-160°
Beilstein Reference: 506001
Merck Reference: 14,3032
UN No.: 2248
MDL No.: MFCD00009429
Formula:
C₈H₁₉N Boiling Pt: 159…160 °C Melting Pt: –62 °C Density: 0.767 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009429 CAS Number: 111-92-2 EINECS: 203-921-8 UN: 2248 ADR: 8,II Merck Index: 14,3032 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...