1,4-Dibromobutane 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dibromo butane

A11669-09 A11669-22 A11669-36
CAAAAA11669-09EA 41.4 CAD
CAAAAA11669-09 CAAAA11669-22 CAAAA11669-36
1,4-Dibromobutane 98+%
1,4-Dibromobutane

MDL: MFCD00000261 Beilstein Registry No.: 1071199


Purity: 98+% CAS No.: 110-52-1 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: 131°(267°F) Refractive Index: 1.5186 Melting Point: -20° Boiling Point: 194-196° Beilstein Reference: 1071199 MDL No.: MFCD00000261

Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 197
Melting Pt: -20
Density: 1.820
Flash Pt: 131 °C (267 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000261
CAS Number: 110-52-1
EINECS: 203-775-5
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR