Semicarbazide hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11668-0E
A11668-22
A11668-36
CAAAA11668-0EEA
483.48
CAD
CAAAA11668-0E
CAAAA11668-22
CAAAA11668-36
Semicarbazide hydrochloride 99%
Semicarbazide hydrochloride
Urease substrate and MAO inhibitor
Purity: 99%
CAS No.: 563-41-7
Molecular Formula: CH4N3O.HCl
Formula Weight: 111.53
Physical Form: Crystalline
Appearance: White
Odor: Odorless
pH: 1.5
Melting Point: ca 176° dec.
Beilstein Reference: 3593642
Merck Reference: 14,8444
UN No.: 2811
MDL No.: MFCD00013009
Urease substrate and MAO inhibitor
Formula:
CH₆ClN₃O Melting Pt: ∼ 176 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013009 CAS Number: 563-41-7 EINECS: 209-247-0 UN: 2811 ADR: 6.1,III Merck Index: 14,8444 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...