2,4-Difluorotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-Difluoro-1-methylbenzene
A11667-06
A11667-14
CAAAA11667-06EA
31.92
CAD
CAAAA11667-06
CAAAA11667-14
2,4-Difluorotoluene 98+%
2,4-Difluorotoluene
MDL: MFCD00000332
Beilstein Registry No.: 1931681
Purity: 98+%
CAS No.: 452-76-6
Molecular Formula: C7H6F2
Formula Weight: 128.12
Flash Point: 15°(59°F)
Refractive Index: 1.4490
Odor: Aromatic
Boiling Point: 114-115°
Beilstein Reference: 1931681
UN No.: 2388
MDL No.: MFCD00000332
Formula:
C₇H₆F₂ MW: 128.12 Boiling Pt: 114 Density: 1.121 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000332 CAS Number: 452-76-6 EINECS: 207-211-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...