Indazole 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1H-Indazole
A11665-03
A11665-06
A11665-14
CAAAA11665-03EA
57.83
CAD
CAAAA11665-03
CAAAA11665-06
CAAAA11665-14
Indazole 99%
Indazole
MDL: MFCD00005691
Beilstein Registry No.: 109676
Fieser: 18,189
Purity: 99%
CAS No.: 271-44-3
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 146-149°
Boiling Point: 270°
Merck Reference: 14,4936
MDL No.: MFCD00005691
Formula:
C₇H₆N₂ MW: 118.14 g/mol Boiling Pt: 270 Melting Pt: 146…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005691 CAS Number: 271-44-3 EINECS: 205-978-4 Merck Index: 14,4936 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...