3,5-Dihydroxybenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Resorcylic acid

A11664-22 A11664-36
CAAAA11664-22EA 88.16 CAD
CAAAA11664-22 CAAAA11664-36
3,5-Dihydroxybenzoic acid 97%
3,5-Dihydroxybenzoic acid

MDL: MFCD00002512 Beilstein Registry No.: 2207864


Purity: 98% CAS No.: 99-10-5 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 237° dec. Beilstein Reference: 2207864

Formula: C₇H₆O₄
MW: 154.12 g/mol
Melting Pt: ∼ 237 °C (dec.)
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002512
CAS Number: 99-10-5
EINECS: 202-730-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR