Caprinaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Decylaldehyde, Decanal, Decyl aldehyde

A11656-AE A11656-AP
CAAAA11656-AEEA 89.59 CAD
CAAAA11656-AE CAAAA11656-AP
Caprinaldehyde 96%
Caprinaldehyde

MDL: MFCD00007031 Beilstein Registry No.: 1362530


Purity: 96% CAS No.: 112-31-2 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 85°(185°F) Refractive Index: 1.4280 Melting Point: -5° Boiling Point: 207-209° Beilstein Reference: 1362530 MDL No.: MFCD00007031

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 207
Melting Pt: 7
Density: 0.828
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007031
CAS Number: 112-31-2
EINECS: 203-957-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR