Calcium-D-gluconate monohydrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate

A11649-0E A11649-22 A11649-36
CAAAA11649-0EEA 235.42 CAD
CAAAA11649-0E CAAAA11649-22 CAAAA11649-36
Calcium-D-gluconate monohydrate 98+%
Calcium D-gluconate monohydrate

MDL: MFCD00064209


Purity: 98+% CAS No.: 66905-23-5 Molecular Formula: C12H22CaO14.H2O Formula Weight: 448.40 (430.37anhy) Physical Form: Powder Appearance: White Odor: Odorless Melting Point: ca 195° dec. Merck Reference: 14,1670 MDL No.: MFCD00064209

Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8
Merck Index: 14,1670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR