Disodium oxalate 99%

Supplier: Thermo Scientific Chemicals
Warning

A11648-0E A11648-22 A11648-36
CAAAA11648-0EEA 526.14 CAD
CAAAA11648-0E CAAAA11648-22 CAAAA11648-36
Disodium oxalate 99%
di-Sodium oxalate

MDL: MFCD00012465 Beilstein Registry No.: 3631622


Purity: 99% CAS No.: 62-76-0 Molecular Formula: Na2C2O4 Formula Weight: 134.00 Odor: Odorless Melting Point: ca 250-270° dec. Beilstein Reference: 3631622 Merck Reference: 14,8650 MDL No.: MFCD00012465

Formula: Na₂C₂O₄
Melting Pt: ∼ 250…270 °C (dec.)
Density: 2.340
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 14,8650

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR