Ethyl myristate 98%
Supplier: Thermo Scientific Chemicals
A11646-AE
A11646-AP
CAAAA11646-AEEA
79.32
CAD
CAAAA11646-AE
CAAAA11646-AP
Ethyl myristate 98%
Ethyl myristate
MDL: MFCD00008984
Beilstein Registry No.: 1776382
Purity: 98%
CAS No.: 124-06-1
Molecular Formula: C16H32O2
Formula Weight: 256.43
Flash Point: 135°(275°F)
Refractive Index: 1.4360
Odor: Weak
Melting Point: 12°
Boiling Point: 295°
Beilstein Reference: 1776382
Merck Reference: 14,6333
MDL No.: MFCD00008984
Formula:
C₁₆H₃₂O₂ Boiling Pt: 295 °C Melting Pt: 12 °C Density: 0.860 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008984 CAS Number: 124-06-1 EINECS: 204-675-4 Merck Index: 14,6333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...