Isopropyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11641-06
A11641-14
CAAAA11641-06EA
71.26
CAD
CAAAA11641-06
CAAAA11641-14
Isopropyl isothiocyanate 97%
Isopropyl isothiocyanate
MDL: MFCD00037830
Beilstein Registry No.: 1739163
Purity: 97%
CAS No.: 2253-73-8
Molecular Formula: (CH3)2CHNCS
Formula Weight: 101.17
Flash Point: 40°(104°F)
Refractive Index: 1.4930
Odor: Pungent
Boiling Point: 137-138°
Beilstein Reference: 1739163
UN No.: 2924
MDL No.: MFCD00037830
Formula:
C₄H₇NS Boiling Pt: 137…138 °C Density: 0.948 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037830 CAS Number: 2253-73-8 EINECS: 218-851-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...