Bronopol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-2-nitro-1,3-propanediol, 2-Bromo-2-nitropropane-1,3-diol

A11639-22 A11639-36
CAAAA11639-22EA 148.52 CAD
CAAAA11639-22 CAAAA11639-36
Bronopol 98+%
Bronopol

MDL: MFCD00007390 Beilstein Registry No.: 1705868


Purity: 98+% CAS No.: 52-51-7 Molecular Formula: C3H6BrNO4 Formula Weight: 199.99 pH: 5-7 Melting Point: ca 122-132° Beilstein Reference: 1705868 Merck Reference: 14,1447 UN No.: 3241 MDL No.: MFCD00007390

Formula: HOCH₂BrC(NO₂)CH₂OH
MW: 199.99 g/mol
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: ∼ 122…132 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007390
CAS Number: 52-51-7
EINECS: 200-143-0
UN: 3241
ADR: 4.1,III
Merck Index: 14,1447

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR