Ethyl-2-methylacetoacetate 95%
Supplier: Thermo Scientific Chemicals
Warning
|
A11629-14
A11629-22
A11629-36
CAAAA11629-14EA
77.1
CAD
CAAAA11629-14
CAAAA11629-22
CAAAA11629-36
Ethyl-2-methylacetoacetate 95%
Ethyl 2-methylacetoacetate
MDL: MFCD00009164
Beilstein Registry No.: 1071742
Purity: 95%
CAS No.: 609-14-3
Molecular Formula: C7H12O3
Formula Weight: 144.17
Flash Point: 62°(143°F)
Refractive Index: 1.4180
Odor: Ester-like
Melting Point: -45°
Boiling Point: 185°
Beilstein Reference: 1071742
MDL No.: MFCD00009164
Formula:
C₇H₁₂O₃ Boiling Pt: 185 °C Melting Pt: –45 °C Density: 1.019 Flash Pt: 62 °C(143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009164 CAS Number: 609-14-3 EINECS: 210-179-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...