2-Fluoropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11627-14
A11627-22
A11627-30
A11627-09
CAAAAA11627-14EA
115.97
CAD
CAAAAA11627-14
CAAAAA11627-22
CAAAAA11627-30
CAAAA11627-09
2-Fluoropyridine 99%
2-Fluoropyridine
MDL: MFCD00006224
Beilstein Registry No.: 1515
Formula:
C₅H₄FN MW: 97.09 g/mol Boiling Pt: 124…126 °C Density: 1.129 Flash Pt: 24 °C (75 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006224 CAS Number: 372-48-5 EINECS: 206-757-5 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...