Pentafluorobenzyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

A11623-22 A11623-06 A11623-14
CAAAAA11623-22EA 865.84 CAD
CAAAAA11623-22 CAAAA11623-06 CAAAA11623-14
Pentafluorobenzyl bromide 98+%
Pentafluorobenzyl bromide

MDL: MFCD00000299 Beilstein Registry No.: 647808


Purity: 98+% CAS No.: 1765-40-8 Molecular Formula: C7H2BrF5 Formula Weight: 261.00 Flash Point: 83°(181°F) Refractive Index: 1.4720 Melting Point: 18-20° Boiling Point: 174-175° Beilstein Reference: 647808 UN No.: 3265 MDL No.: MFCD00000299

Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
Boiling Pt: 174…175 °C
Melting Pt: 18…20 °C
Density: 1.844
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR