3-Methylveratrole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Dimethoxy-3-methylbenzene, 2,3-Dimethoxytoluene
A11617-06
A11617-14
CAAAA11617-06EA
65.89
CAD
CAAAA11617-06
CAAAA11617-14
3-Methylveratrole 98+%
3-Methylveratrole
MDL: MFCD00008379
Beilstein Registry No.: 1940962
Purity: 98+%
CAS No.: 4463-33-6
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 85°(185°F)
Refractive Index: 1.5140
Odor: Aromatic
Boiling Point: 202-203°
Beilstein Reference: 1940962
MDL No.: MFCD00008379
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 202…203 °C Density: 1.024 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008379 CAS Number: 4463-33-6 EINECS: 224-726-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...