Potassium benzoate 99%
Supplier: Thermo Scientific Chemicals
A11610-0B
A11610-30
CAAAA11610-0BEA
195.92
CAD
CAAAA11610-0B
CAAAA11610-30
Potassium benzoate 99%
Potassium benzoate
MDL: MFCD00013061
Beilstein Registry No.: 3719165
Purity: 99%
CAS No.: 582-25-2
Molecular Formula: C7H5KO2
Formula Weight: 160.22
Physical Form: Powder
Sensitivity: Hygroscopic
Beilstein Reference: 3719165
Merck Reference: 14,1091
MDL No.: MFCD00013061
Formula:
C₇H₅KO₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013061 CAS Number: 582-25-2 EINECS: 209-481-3 Merck Index: 14,1091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...