Triphenylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11605-09
A11605-18
CAAAA11605-09EA
67.15
CAD
CAAAA11605-09
CAAAA11605-18
Triphenylsilane 97%
Triphenylsilane
MDL: MFCD00003003
Beilstein Registry No.: 978182
Purity: 97%
CAS No.: 789-25-3
Molecular Formula: (C6H5)3SiH
Formula Weight: 260.41
Flash Point: 76°(169°F)
Melting Point: 42-44°
Boiling Point: 159-163°/3mm
Beilstein Reference: 978182
MDL No.: MFCD00003003
Formula:
C₁₈H₁₆Si Boiling Pt: 159…163 °C/3 mm Melting Pt: 42…44 °C Density: 1.152 g/cm³ (20 °C) Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003003 CAS Number: 789-25-3 EINECS: 212-333-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...