Phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11596-06
A11596-22
A11596-36
CAAAAA11596-06EA
52.77
CAD
CAAAAA11596-06
CAAAA11596-22
CAAAA11596-36
Phenyl isothiocyanate 97%
Phenyl isothiocyanate
MDL: MFCD00004798
Beilstein Registry No.: 471392
Fieser: 1,844 2,323
Purity: 97%
CAS No.: 103-72-0
Molecular Formula: C7H5NS
Formula Weight: 135.19
Flash Point: 87°(188°F)
Refractive Index: 1.6500
Odor: Pungent
Melting Point: -21°
Boiling Point: 220-221°
Beilstein Reference: 471392
Merck Reference: 14,7297
UN No.: 2927
MDL No.: MFCD00004798
Formula:
C₇H₅NS Boiling Pt: 220…221 °C Melting Pt: –21 °C Density: 1.131 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004798 CAS Number: 103-72-0 EINECS: 203-138-1 UN: 2927 ADR: 6.1,II Merck Index: 14,7297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...