Ethylene glycol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

A11591-0C A11591-0E A11591-30 A11591-36
CAAAAA11591-0CEA 578.28 CAD
CAAAAA11591-0C CAAAAA11591-0E CAAAAA11591-30 CAAAAA11591-36
Ethylene glycol 99%
Ethylene glycol

MDL: MFCD00002885 Beilstein Registry No.: 505945 Fieser: 1,375 5,296 9,217 15,156 18,157


Purity: 99% CAS No.: 107-21-1 Molecular Formula: C2H6O2 Formula Weight: 62.07 Flash Point: 119°(246°F) Refractive Index: 1.4310 Odor: Sweet Melting Point: -13° Boiling Point: 196-198° Beilstein Reference: 505945 Merck Reference: 14,3798 MDL No.: MFCD00002885

Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C
Melting Pt: –13 °C
Density: 1.113
Flash Pt: 119 °C(246 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 14,3798
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR