4-Methylanisole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11572-0F
A11572-AP
CAAAA11572-0FEA
434.5
CAD
CAAAA11572-0F
CAAAA11572-AP
4-Methylanisole 99%
4-Methylanisole
MDL: MFCD00008413
Beilstein Registry No.: 1237336
Purity: 99%
CAS No.: 104-93-8
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 58°(136°F)
Refractive Index: 1.5110
Odor: Aromatic
Melting Point: -50°
Boiling Point: 174-175°
Beilstein Reference: 1237336
UN No.: 3271
MDL No.: MFCD00008413
Formula:
C₈H₁₀O Boiling Pt: 174…175 °C Melting Pt: –50 °C Density: 0.968 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008413 CAS Number: 104-93-8 EINECS: 203-253-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...